Learn More
ATCC COLO 201; Colon Cancer; Human (Homo sapiens)
Catalog No. 502384897
Encompass
COLO-201 is a bipolar, slightly refractile, fibroblast-like cell line that was isolated from the colon of a White, 70-year-old, male patient with colorectal adenocarcinoma. This cell line was deposited by GE Moore and can be used in toxicology and cancer research.
Catalog No. 50-238-4897 Supplier ATCC Supplier No. CCL224
COLO-201 is a bipolar, slightly refractile, fibroblast-like cell line that was isolated from the colon of a White, 70-year-old, male patient with colorectal adenocarcinoma. This cell line was deposited by GE Moore and can be used in toxicology and cancer research.