Learn More
ATCC COLO 205; Colon Adenocarcinoma; Human (Homo sapiens)
Catalog No. 502384892
Encompass
The COLO 205 cell line is made up of epithelial cells isolated in 1975 from ascitic fluid derived from a 70-year-old, White, male with colon cancer. The cells can be used for cancer and toxicology research.
Catalog No. 50-238-4892 Supplier ATCC Supplier No. CCL222
The COLO 205 cell line is made up of epithelial cells isolated in 1975 from ascitic fluid derived from a 70-year-old, White, male with colon cancer. The cells can be used for cancer and toxicology research.