Learn More
US Biomax Inc Colon cancer, colonic disease and normal colon control tissue array, including pathology grade, with MMR (MLH1/PMS2/MSH2/MSH6) IHC results, TNM and clinical stage(AJCC 8th edition), 102 cases/102 cores (cores size 1.5mm)
SDP
Supplier:  US Biomax Inc CO1021C
Encompass
Colon adenocarcinoma with colon tissue microarray, containing 87 cases of adenocarcinoma, 3 papillary adenocarcinoma, 7 mucinous adenocarcinoma, 2 adenoma, 2 colon tissue and 1 polyp of colon, single core per case
Catalog No. NC3789251