Learn More
Hach Company Colony Counter, Quebec, 220 Vac, 50 Hz
Supplier:  Hach Company 2252102
Encompass
For counting bacterial colonies. Features 11.5 cm (4.5") lens with 1.5x magnification to eliminate parallax erros; 40-watt tungsten bulbs to illuminate dark backgrounds; Wolffhuegel guide plate and 3-wire cord. Accomodates Stewart plates. Sheet metal case. Dimensions: 25 x 28 x 27 cm (10 x 11x 10.5"). CSA certified. 220V
Catalog No. 50-199-5637