Learn More
Bel-Art™ Colony Counter System with Mini LED Light Box and Magnifier
Catalog No. 07910025 Shop All SP Bel-Art & SP Wilmad Products
Change view
Click to view available options
Display Type:
None
Catalog No. Display Type
07-910-025 None
1 options
Catalog No. 07-910-025 Supplier Bel-Art™ Supplier No. F378633000
Get Up to $500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
Complete, effective and accurate way to count colonies of bacteria or other microorganisms grown on an agar plate
  • Provides equipment for proper lighting, magnification and tracking of colonies
Includes:
  • Hand-held colony counter (F37862-0002)
    • Marks, counts and gives audible and visual alert each time count is registered
  • Mini LED light box (F37864-3000)
    • Enhances viewing of transparent and semi-transparent objects
  • Mini magnifier (F37865-0000)
    • 1.75X magnification
Specifications
Product Type Colony Counter System
Display Type None
For Use With (Application) For counting colonies of bacteria or other microorganisms grown on an agar plate
Includes Colony Counter System, Mini LED Light Box, Magnifier