Learn More
Biomatik Corporation Colony Stimulating Factor 3, Granulocyte (Gcsf) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU28877
Encompass_Preferred
Colony Stimulating Factor 3, Granulocyte (Gcsf) Polyclonal Antibody is a Pig (Sus scrofa; Porcine) Polyclonal Antibody to Colony Stimulating Factor 3, Granulocyte (GCSF). The Colony Stimulating Factor 3, Granulocyte (Gcsf) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Pig (Sus scrofa; Porcine). Immunogen: Phe34~Ala193. Applications: WB, IHC, ICC, IP. Target Synonyms: Colony Stimulating Factor 3, Granulocyte (GCSF). Accession Number: O02837. Gene ID: 396839. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5313