Learn More
World Precision Instrument Color CCD Video Camera-COLCAM-HD1080P
SDP
Supplier:  World Precision Instrument COLCAMHD1080P
Encompass
Record live video images
The COLCAM-HD1080P is a CMOS-based camera that outputs a true HD 1080P or 720P image at 60fps in the 16x9 format. The camera features the capability to program individual DSP profiles accessed via remote hand held controller.
There is no software or a computer interface. To connect to a computer for viewing or recording via USB requires an external video digitizer. Contact your WPI representative for details.
Catalog No. 50-253-8886