Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vikan™ Color-Coded Scraper, 3 in.

Catalog No. 17227693 Shop All Remco Products
Change view
Click to view available options
Color:
Black
Blue
Green
Orange
Pink
Purple
White
Yellow
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-227-693 White
17-227-695 Orange
17-227-694 Yellow
17-227-691 Green
17-227-692 Blue
17-227-696 Purple
17-227-690 Pink
17-227-697 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 17-227-693 Supplier Vikan™ Supplier No. 69614
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Color-coded scraper supports food safety material handling, cleaning, and sanitation programs that are aimed at preventing cross-contamination.

  • Ideal solution to your bench top scraping needs
  • Has no seams or cracks, which helps prevent bacterial growth

Specifications

Product Type Scraper
Color-coded Yes
Sterility Non-sterile
Certifications/Compliance FDA Compliant
Color White
Dimensions (L x W) 0.75 x 3 in.
Material Polypropylene
Quantity 1 Ea.
Size Medium
Weight (English) 0.1 lb.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.