Learn More
Electron Microscopy Sciences Tough-Tag Color Coded Dual Labels, White
SDP
Supplier:  Electron Microscopy Sciences 77539W
Encompass_Preferred
Color Coded Tough-Tags will not come off in rotors, cyclers or racks. They are resistant to extreme temperatures and the matte surface is suitable for pencil, pen or marker. Ideal for 1.5 ml tubes. Label size: 1.25" x 0.5625" with 0.44" spot (32 x 14 mm with 11 mm spot)
Color: White
Catalog No. 50-337-42