Learn More
Description
- Designed for use with Remco™ Transport Storage Tubs
- They are manufactured from high-density polyethylene
- Suitable for transporting, freezing, and storing
- This model has an angled edge that fits securely on angled tubs
- These lids meet all USDA specifications
- Commonly used for meat, seafood, poultry, commercial fishing, confectionery, bakery, and grain applications
Specifications
Specifications
| Product Type | Tub Lid |
| Color | Red |
| Material | Polyethylene |
| For Use With (Equipment) | Transport Storage Tub |
| Certifications/Compliance | FDA Compliant |
| Quantity | 1 |
| Dimensions (L x W x H) | 46.9 x 26.9 x 1.7 in. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.