Learn More
Hach Company pH, Bromthymol Blue, Color Disc, 5.4-8.4
Supplier:  Hach Company 9261200
Encompass
Color disc for pH, Bromthymol Blue, 5.4-8.4. Used in test kit Models AD-16C, AD-16D, AD-16E, and 17-F.
Catalog No. 50-199-7287