Learn More
Hach Company Iron Ferrozine Color Disc, Low Range, Indoor Light, 2 pcs.
Supplier:  Hach Company 9264400
Encompass
Color disc, used for measurement of Iron in Kit 255600. Range 0.0-0.20 mg Fe/L. Smallest increment, 0.01 mg Fe/L.
Catalog No. 50-199-7310