Learn More
Electron Microscopy Sciences Lab-Tape™ Color Tape, 0.5" wide, Blue 180 FT
SDP
Supplier:  Electron Microscopy Sciences 7743101BL
Encompass_Preferred
For general laboratory use. Easy to write on with permanent markers. Water-resistant. Can be used on small diameter vials.
Width: 0.5"
Color: Blue
Catalog No. 50-193-0465