Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Color Reference Solution Acc. to APHA, Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111003661 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
20 x 10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-3661 20 x 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-3661 Supplier MilliporeSigma Supelco Supplier No. 771471SETF
Only null left
Add to Cart
Add to Cart

contains Cobalt(II) chloride

TRUSTED_SUSTAINABILITY

Specifications

Description This is a set of the following color reference solutions according to APAH:. Colour Reference Solution 5 - 10 mL ampule. Colour Reference Solution 10 - 10 mL ampule. Colour Reference Solution 15 - 10 mL ampule. Colour Reference Solution 20 - 10 mL ampule. Colour Reference Solution 25 - 10 mL ampule. Colour Reference Solution 30 - 10 mL ampule. Colour Reference Solution 35 - 10 mL ampule. Colour Reference Solution 40 - 10 mL ampule. Colour Reference Solution 50 - 10 mL ampule. Colour Reference Solution 60 - 10 mL ampule. Colour Reference Solution 70 - 10 mL ampule. Colour Reference Solution 100 - 10 mL ampule. Colour Reference Solution 150 - 10 mL ampule. Colour Reference Solution 200 - 10 mL ampule. Colour Reference Solution 250 - 10 mL ampule. Colour Reference Solution 300 - 10 mL ampule. Colour Reference Solution 350 - 10 mL ampule. Colour Reference Solution 400 - 10 mL ampule. Colour Reference Solution 450 - 10 mL ampule. Colour Reference Solution 500 - 10 mL ampule
Quantity 20 x 10 mL
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.