Learn More
Hach Company Hach CL17sc Colorimetric Chlorine Analyzer with Standpipe Installation Kit and Reagents for Free Chlorine
Supplier: Hach Company 8572700
The CL17sc colorimetric chlorine analyzer provides continuous, EPA-approved DPD measurement of free chlorine for water quality monitoring. It reduces routine maintenance with simplified reagent handling, offers user-friendly diagnostics for quick troubleshooting, and supports integration with controller platforms for centralized data collection and remote monitoring.
- Continuous colorimetric DPD measurement of free chlorine.
- Measurement range 0.03-10 mg/L Cl₂ for broad monitoring needs.
- Compact enclosure with simplified tubing replacement to reduce downtime.
- Built-in flow indication and LED status lights for quick operational checks.
- Includes standpipe installation kit and a supply of reagents to begin operation.
- Designed for integration with external controllers and SCADA systems.