Learn More
Perkin Elmer US LLC Calibrated Diffuse Color Standards - 2 in, Qty. 4
Supplier:  Perkin Elmer US LLC PELA9021
Encompass_Preferred
Colors for this set are orange, purple, cyan and violet. This set of 4 calibrated 2 inch diameter diffuse color standards are used for establishment of photometric accuracy and linearity in diffuse reflectance mode. Calibration is traceable to NIST and NPL standards, and is calibrated 250-2500 nm reported each 50 nm. Disc is included with data each 1 nm.
Catalog No. 50-201-6541
Add to Cart
Request a Quote