Learn More
Sigma Aldrich Fine Chemicals Biosciences Aldrich ColorSpec NMR tubes, 5 mm x 8 in, 50/pk
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z56920850EA
Encompass
This new range of premium quality NMR sample tubes feature color-coded banding and caps and a MHz rating that allows instant visual identification to tube grade and application. No more mix ups after cleaning. Concentricity × camber:102 μm × 102 μm; Parameter:200 MHz frequency; Color:color-coded blue
Catalog No. NC0861849