Learn More
Helena Laboratories™ ColoScreen™ FOB Test Kit
SDP
Catalog No. 23038049
Change view
Click to view available options
Quantity:
100 Pack
Type:
ColoScreen Lab Pack
ColoScreen Office Pack
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
23-038-049 100 Pack ColoScreen Lab Pack
23-038-046 100 Pack ColoScreen Office Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-038-049 Supplier Helena Laboratories™ Supplier No. 5082
Add to Cart
Add to Cart
Convenient triple slide provides three successive tests in one unit
  • CLIA Complexity: Waived
  • Ideal for physician's office or clinic use
  • Classic guaiac slide format
  • Office Pack allows slides, sample applicators and mailing envelopes to be conveniently distributed to patients, who can then prepare test slides in the privacy of their homes
  • Clinic Pack is designed for labs preferring the triple unit slides
  • Lab Pack is provided in order that each laboratory can stock the volume that best fits its demands
  • Each single slide in the triple unit has two test areas so that multiple sites from each specimen may be tested, increasing accuracy
Specifications
Clia Complexity Waived
Type ColoScreen Lab Pack
For Use With (Application) Fecal Occult Blood Testing
Includes 34 Triple unit ColoScreen slides with on-slide, ColoCheck monitors, 2 x 15mL ColoScreen developer, 100 specimen applicator
Quantity 100 Pack