Learn More
PCE AMERICAS INC Colorimeter features a measuring aperture of 8 mm.
SDP
Catalog No. NC3654622
Encompass_Preferred
"A very precise Colorimeter with 45°/0° viewing angle. This very professional instrument works in color spaces CIEL*a*b*C*h*, CIEL*a*b*, and CIEXYZ. This kind of color meter is mainly made for the use in an R&D laboratory, for industrial product evaluation, or for any kind of color testing application. The Colorimeter comes fully equipped (click on detail sheet) and is manufacturer calibrated. - A color meter for industrial professionals and researchers -Measuring aperture of 8 mm and 45°/0° illumination/viewing angleRechargeable lithium-ion batteryAuto white black calibrationDesigned specifically for the food industrySoftware IncludedStores 100 reference values and 20,000 samples"
Catalog No. NC3654622 Supplier PCE AMERICAS INC Supplier No. PCECSM 2
"A very precise Colorimeter with 45°/0° viewing angle. This very professional instrument works in color spaces CIEL*a*b*C*h*, CIEL*a*b*, and CIEXYZ. This kind of color meter is mainly made for the use in an R&D laboratory, for industrial product evaluation, or for any kind of color testing application. The Colorimeter comes fully equipped (click on detail sheet) and is manufacturer calibrated. - A color meter for industrial professionals and researchers -Measuring aperture of 8 mm and 45°/0° illumination/viewing angleRechargeable lithium-ion batteryAuto white black calibrationDesigned specifically for the food industrySoftware IncludedStores 100 reference values and 20,000 samples"