Learn More
PCE AMERICAS INC Colorimeter features a measuring geometry 8°/d.
SDP
Catalog No. NC3654874
Encompass_Preferred
"The color meter PCE-CSM 3 was developed for quality control and offers a very high accuracy. When using the color meter, you can choose between different color spaces (CIE L*a*b*C*h, CIE L*a*b*). After a measurement with the color meter, the reading can be used as a reference value while other readings can be recorded as a sample and later be compared to the reference value easily.  In addition to the individual color space coordinates, the deviation between the reading is also displayed. When using the color meter, it is possible to store up to 100 reference values and up to 20,000 samples on the device. The measured data can also be transferred to a PC via the USB interface. A special PC software is included in the package."
Catalog No. NC3654874 Supplier PCE AMERICAS INC Supplier No. PCECSM 3
"The color meter PCE-CSM 3 was developed for quality control and offers a very high accuracy. When using the color meter, you can choose between different color spaces (CIE L*a*b*C*h, CIE L*a*b*). After a measurement with the color meter, the reading can be used as a reference value while other readings can be recorded as a sample and later be compared to the reference value easily.  In addition to the individual color space coordinates, the deviation between the reading is also displayed. When using the color meter, it is possible to store up to 100 reference values and up to 20,000 samples on the device. The measured data can also be transferred to a PC via the USB interface. A special PC software is included in the package."