Learn More
PCE AMERICAS INC Colorimeter features a measuring aperture Ø 4 mm / Ø 8 mm
SDP
Catalog No. NC3654880
Encompass_Preferred
"PCE-CSM 7 is a portable colorimeter used to measure different spectral parameters. This user-friendly, ergonomically designed colorimeter automatically performs a white and black calibration when powered on. Suitable for manufacturing quality control and assurance as well as product research and development, the colorimeter offers impressive measuring accuracy and comes manufacturer calibrated. Camera/illumination locating for small samples.Color spaces: CIEL*a*b*C*h* CIEL*a*b* CIEXYZ CIERGB CIEL*u*v* CIEL*C*h +Yellowness + Whiteness + Color FastnessOffers two different measuring aperturesThree light sources D65, D50, Aoptional liquid sample kit listed under accessoriesStorage of 100 reference values and 20,000 samplesIllumination/Viewing Angles 8°/d "
Catalog No. NC3654880 Supplier PCE AMERICAS INC Supplier No. PCECSM 7
Add to Cart
Add to Cart
Request a Quote
"PCE-CSM 7 is a portable colorimeter used to measure different spectral parameters. This user-friendly, ergonomically designed colorimeter automatically performs a white and black calibration when powered on. Suitable for manufacturing quality control and assurance as well as product research and development, the colorimeter offers impressive measuring accuracy and comes manufacturer calibrated. Camera/illumination locating for small samples.Color spaces: CIEL*a*b*C*h* CIEL*a*b* CIEXYZ CIERGB CIEL*u*v* CIEL*C*h +Yellowness + Whiteness + Color FastnessOffers two different measuring aperturesThree light sources D65, D50, Aoptional liquid sample kit listed under accessoriesStorage of 100 reference values and 20,000 samplesIllumination/Viewing Angles 8°/d "