Learn More
Aviva Systems Biology Corp COLQ Antibody - N-terminal region (ARP34362_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34362_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against COLQ. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human COLQ. Alternate names: EADApplicationWB. Species reactivity: Cow, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53814