Learn More
Thermo Scientific Dionex IonPac AS22-Fast-4m IC Columns | | 1 EA
Catalog No. NC0996047 Shop All Thermo Scientific Products
Encompass
Fast separation of common inorganic anions with excellent resolution,Obtain the highest peak efficiency through the use of smaller resin particles of common inorganic anions in diverse sample matrices,Attain fast isocratic separation of the common inorganic anions in less than 5 min,Perform compliance monitoring of common anions in drinking water as specified in US EPA Method 3000 A,Achieve excellent separation of inorganic anions and low-molecular weight acids in diverse sample matrices,Obtain high peak efficiency and increased peak resolution for more consistent integration and more reliable results,Transfer application easily
Catalog No. NC0996047 Supplier Thermo Scientific Supplier No. 088486
Fast separation of common inorganic anions with excellent resolution,Obtain the highest peak efficiency through the use of smaller resin particles of common inorganic anions in diverse sample matrices,Attain fast isocratic separation of the common inorganic anions in less than 5 min,Perform compliance monitoring of common anions in drinking water as specified in US EPA Method 3000 A,Achieve excellent separation of inorganic anions and low-molecular weight acids in diverse sample matrices,Obtain high peak efficiency and increased peak resolution for more consistent integration and more reliable results,Transfer application easily