Learn More
Medchemexpress LLC Columbamine chloride | 1916-10-5 | 373.83 g/mol | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN0926A5MG
Encompass_Preferred
Columbamine chloride (Synonyms: Columbamin chloride; Dehydroisocorypalmine chloride) is an organic heterotetracyclic alkaloid extracted from plants. Columbamine chloride is a metabolite of Berberine. Columbamine chloride inhibits the cytochrome P450 (CYP) isoform CYP3A4 (IC50 = 30.6 μM). Columbamine chloride induces apoptosis in cancer cells. Columbamine chloride can be used for antioxidant, anti-inflammatory, antitumor, antifungal, antiparasite, hepatoprotective, neuroprotective, hypolipidemic and hypoglycemic study.
  • Organic heterotetracyclic alkaloid extracted from plants.
  • Metabolite of Berberine.
  • Inhibits the cytochrome P450 (CYP) isoform CYP3A4.
  • Induces apoptosis in cancer cells.
  • Used for antioxidant, anti-inflammatory, antitumor, antifungal, antiparasite, hepatoprotective, neuroprotective, hypolipidemic, and hypoglycemic study.
Catalog No. 50-004-16991