Learn More
MOLTOX Molecular Toxicology Inc Columbia Agar, 20 mL/plate, 10 plates/sleeve, Store at Room Temperature
SDP
Catalog No. 50948623
Encompass_Preferred
This general purpose medium is used for the isolation and cultivation of fastidious and nonfastidious microorganisms. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-623 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 2140S111
This general purpose medium is used for the isolation and cultivation of fastidious and nonfastidious microorganisms. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.