Learn More
Bioworld Columbia Blood Agar Base, 1 kg
Catalog No. 50254296
Encompass_Preferred
Columbia Blood Agar BaseUsed for the cultivation and determining of hemolytic reactions of fastidious and non-fastidious microorganismsSupplemented with 5-10% sheep, rabbit, or horse bloodpH: 7.3Dehydrated Appearance: beige, homogeneous powderPrepared Appearance: without blood- light to medium amber agar with 5% sheep blood- red agar Soluble in boiling waterContains:Pancreatic Digest of Casein Peptic Digest of Animal Tissue Yeast enriched Peptone Corn Starch Sodium Chloride Agar
Catalog No. 50-254-296 Supplier Bioworld Supplier No. 306200202
Columbia Blood Agar BaseUsed for the cultivation and determining of hemolytic reactions of fastidious and non-fastidious microorganismsSupplemented with 5-10% sheep, rabbit, or horse bloodpH: 7.3Dehydrated Appearance: beige, homogeneous powderPrepared Appearance: without blood- light to medium amber agar with 5% sheep blood- red agar Soluble in boiling waterContains:Pancreatic Digest of Casein Peptic Digest of Animal Tissue Yeast enriched Peptone Corn Starch Sodium Chloride Agar