Learn More
Biolog Inc CA - Columbia Blood Agar, 4 Plates/Pkg
SDP
Catalog No. NC1446647
Encompass
This item has a minimum qty of 25 per supplier requirements.
Columbia Blood Agar (CA) is intended for the isolation, quantitation, and partial identification of a variety of microorganisms, including fastidious organisms from clinical specimens. This media is prepared, stored, and dispensed under oxygen-free conditions to prevent the formation of oxidized products prior to use.CA is suitable for use in antibiotic differential disk examination and spot biochemical testing. CA is an enriched non-selective media. It is supplemented with vitamin K1 and hemin to facilitate the recovery of fastidious anaerobic bacteria. Sheep blood is added for the observation of hemolytic reactions as seen by the double zone -hemolysis of Clostridium perfringens, and for growth factors required by some fastidious microorganisms.
This item is not returnable. View return policy
Catalog No. NC1446647 Supplier Biolog Inc Supplier No. AS895
This item has a minimum qty of 25 per supplier requirements.
Columbia Blood Agar (CA) is intended for the isolation, quantitation, and partial identification of a variety of microorganisms, including fastidious organisms from clinical specimens. This media is prepared, stored, and dispensed under oxygen-free conditions to prevent the formation of oxidized products prior to use.CA is suitable for use in antibiotic differential disk examination and spot biochemical testing. CA is an enriched non-selective media. It is supplemented with vitamin K1 and hemin to facilitate the recovery of fastidious anaerobic bacteria. Sheep blood is added for the observation of hemolytic reactions as seen by the double zone -hemolysis of Clostridium perfringens, and for growth factors required by some fastidious microorganisms.