Learn More
Bioworld Columbia Broth, 500 g
Catalog No. 50254298
Encompass_Preferred
Columbia BrothUsed for the cultivation of fastidious and non-fastidious microorganisms from clinical specimensAn excellent blood culture medium when in the presence of CO2 and supplemented with SPSpH: 7.5Prepared Appearance: light amber brothContains:Pancreatic Digest of Casein Peptic Digest of Animal Tissue Yeast Enriched Peptone Sodium Chloride Dextrose L-Cysteine-HC Magnesium Sulfate Ferrous Sulfate Tris (Hydroxymethylaminomethane) Tris-HCI Sodium Carbonate
Catalog No. 50-254-298 Supplier Bioworld Supplier No. 306200211
Columbia BrothUsed for the cultivation of fastidious and non-fastidious microorganisms from clinical specimensAn excellent blood culture medium when in the presence of CO2 and supplemented with SPSpH: 7.5Prepared Appearance: light amber brothContains:Pancreatic Digest of Casein Peptic Digest of Animal Tissue Yeast Enriched Peptone Sodium Chloride Dextrose L-Cysteine-HC Magnesium Sulfate Ferrous Sulfate Tris (Hydroxymethylaminomethane) Tris-HCI Sodium Carbonate