Learn More
Research Products International Corp Columbia CNA Agar, 1 Kilogram
SDP
Catalog No. 501368162
Encompass_Preferred
Selective Agar medium used for the isolation of Gram positive staphylococci and streptococci from clinical specimens. Eliner et al. added colistin nalidixic acid to Columbia Agar to suppress the growth of gram negative bacteria. The addition of 5-10% sheep red blood cells to the base demonstrates hemolytic patterns of the staphylococci and streptococci.
Formula per liter*:Casein Digest Peptone: 5.0g; Corn Starch: 1.0g; Peptic Digest of Animal Tissue: 8.0g; Sodium Chloride: 5.0g; Yeast Enriched Peptone: 10.0g; Colistine: 0.015g; Nalidixic Acid: 0.010g; Agar: 14.0g. *Grams per liter may be adjusted or formula supplemented to obtain desired performance.
Preparation: Mix 44 grams of the medium in a liter of purified water until evenly dispersed. Heat with repeated stirring and boil for one minute to dissolve completely. Distribute and autoclave at 121°C for 15 minutes.
Catalog No. 50-136-8162 Supplier Research Products International Corp Supplier No. C400501000.0
Selective Agar medium used for the isolation of Gram positive staphylococci and streptococci from clinical specimens. Eliner et al. added colistin nalidixic acid to Columbia Agar to suppress the growth of gram negative bacteria. The addition of 5-10% sheep red blood cells to the base demonstrates hemolytic patterns of the staphylococci and streptococci.
Formula per liter*:Casein Digest Peptone: 5.0g; Corn Starch: 1.0g; Peptic Digest of Animal Tissue: 8.0g; Sodium Chloride: 5.0g; Yeast Enriched Peptone: 10.0g; Colistine: 0.015g; Nalidixic Acid: 0.010g; Agar: 14.0g. *Grams per liter may be adjusted or formula supplemented to obtain desired performance.
Preparation: Mix 44 grams of the medium in a liter of purified water until evenly dispersed. Heat with repeated stirring and boil for one minute to dissolve completely. Distribute and autoclave at 121°C for 15 minutes.