Learn More
NEOGEN Columbia CNA Agar, 10 kg
SDP
Catalog No. 502015485
Encompass_Preferred
Columbia CNA Agar is used with blood for the selective isolation of Gram-positive cocci in a laboratory setting. Columbia CNA Agar is not intended for use in the diagnosis of disease or other conditions in humans.Ellner et al. described Columbia CNA Agar as a variation of Columbia Blood Agar Base that is selective for Gram-positive cocci. Colistin and Nalidixic Acid are added to the formula, selecting for Gram-positive organisms and fungi by suppressing Gram-negative bacteria.For more information visit https://www.neogen.com/solutions/Microbiology/columbia-cna-agar
Catalog No. 50-201-5485 Supplier NEOGEN Supplier No. 700003298
Columbia CNA Agar is used with blood for the selective isolation of Gram-positive cocci in a laboratory setting. Columbia CNA Agar is not intended for use in the diagnosis of disease or other conditions in humans.Ellner et al. described Columbia CNA Agar as a variation of Columbia Blood Agar Base that is selective for Gram-positive cocci. Colistin and Nalidixic Acid are added to the formula, selecting for Gram-positive organisms and fungi by suppressing Gram-negative bacteria.For more information visit https://www.neogen.com/solutions/Microbiology/columbia-cna-agar