Learn More
Thermo Scientific™ Columbia CNA Agar with Sheep Blood Medium
Description
Effectively isolate Gram-positive organisms from clinical samples using Thermo Scientific™ Columbia CNA Agar with Sheep Blood, a selective microbiological medium that includes colistin and nalidixic acid to inhibit gram-negative bacilli. Columbia CNA Agar has traditionally been used to identify staphylococci and streptococci. The addition of sheep blood to the medium allows distinct identification of S. pneumoniae through the production of clear alpha-haemolysis.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Specifications
| Color | Opaque red |
| Content And Storage | 2°C to 8°C |
| Format | 85mm Monoplate |
| Manufacturing Origin | United States |
| Media Type | CNA with 5% Sheep Blood |
| Product Type | Agar Plate |
| Fill Volume | 18 mL |
| Includes | Colistin, Nalidixic Acid |
| Packaging Type | Flow Wrapped - Single Layer |
| Quantity | 10/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.