Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Columbia CNA Agar with Sheep Blood Medium

Catalog No. r01322
Change view
Click to view available options
Quantity:
10/Pk.
100/Pk.
15/Pk.
Format:
10mm x 100mm Spacesaver Plate
85mm Monoplate
Media Type:
CNA
CNA with 5% Sheep Blood
Includes:
Colistin, Nalidixic Acid
Nalidixic Acid
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format Media Type Includes
R01322 100/Pk. 85mm Monoplate CNA with 5% Sheep Blood Colistin, Nalidixic Acid
R01320 10/Pk. 85mm Monoplate CNA with 5% Sheep Blood Colistin, Nalidixic Acid
R01321 15/Pk. 10mm x 100mm Spacesaver Plate CNA Nalidixic Acid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. R01322 Supplier Thermo Scientific™ Supplier No. R01322
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Isolate Gram-positive organisms from clinical samples with this selective media traditionally used to identify staphylococci, streptococci, and S. pneumoniae.

Effectively isolate Gram-positive organisms from clinical samples using Thermo Scientific™ Columbia CNA Agar with Sheep Blood, a selective microbiological medium that includes colistin and nalidixic acid to inhibit gram-negative bacilli. Columbia CNA Agar has traditionally been used to identify staphylococci and streptococci. The addition of sheep blood to the medium allows distinct identification of S. pneumoniae through the production of clear alpha-haemolysis.

Columbia CNA Agar with Sheep Blood contains antibiotics to inhibit S. albus and Micrococcus species as well as Gram-positive and Gram-negative rods. It suppresses growth of Proteus, Klebsiella and Pseudomonas species while permitting unrestricted growth of S. aureus, haemolytic streptococci and enterococci. This medium enables important Gram-positive cocci to be recognised more readily and isolated easily from the mixed bacterial populations contained in many clinical specimens and foods.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY

Specifications

Color Opaque red
Content And Storage 2°C to 8°C
Format 85mm Monoplate
Manufacturing Origin United States
Media Type CNA with 5% Sheep Blood
Product Type Agar
Fill Volume 18 mL
Includes Colistin, Nalidixic Acid
Packaging Type Flow Wrapped - Single Layer
Quantity 100/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.