Learn More
Ted Pella Inc COLUMBIA JAR FOR COVERSLIP
SDP
Catalog No. NC1804007
Encompass
This staining jar can hold up to 4 cover slips measuring 17-23mm wide, and up to 30mm long. Longer coverslips can be accommodated by removing the cap. The jar comes with a white polypropylene screw cap with a PTFE coated polyethylene liner.
Catalog No. NC1804007 Supplier Ted Pella Inc Supplier No. 21040
This staining jar can hold up to 4 cover slips measuring 17-23mm wide, and up to 30mm long. Longer coverslips can be accommodated by removing the cap. The jar comes with a white polypropylene screw cap with a PTFE coated polyethylene liner.