Learn More
Medchemexpress LLC (+)-Columbianetin | 3804-70-4 | 246.26 | 5 MG
SDP
Catalog No. 5000389692
Encompass_Preferred
(+)-Columbianetin is an active compound that functions as an inhibitor of JNK and ERK, and an activator of TGFβ signaling. It effectively mitigates UVA-induced cellular damage by reducing phosphorylation of JNK and ERK, decreasing MMP-1 production, and reversing collagen degradation. Additionally, it provides protection against UVB-induced harm to human keratinocytes.
  • Inhibits JNK and ERK pathways.
  • Activates TGFβ signaling.
  • Reduces MMP-1 production.
  • Reverses UVA-induced collagen degradation.
  • Alleviates UVA-mediated inhibition of Smad2/3 phosphorylation.
  • Regulates AP-1 and ASK1-MAPK signaling pathways.
  • Inhibits ROS production.
  • Protects human keratinocytes against UVB-induced damage.
  • Applicable for research related to skin aging.
Catalog No. 50-003-89692 Supplier Medchemexpress LLC Supplier No. HYN03635MG
(+)-Columbianetin is an active compound that functions as an inhibitor of JNK and ERK, and an activator of TGFβ signaling. It effectively mitigates UVA-induced cellular damage by reducing phosphorylation of JNK and ERK, decreasing MMP-1 production, and reversing collagen degradation. Additionally, it provides protection against UVB-induced harm to human keratinocytes.
  • Inhibits JNK and ERK pathways.
  • Activates TGFβ signaling.
  • Reduces MMP-1 production.
  • Reverses UVA-induced collagen degradation.
  • Alleviates UVA-mediated inhibition of Smad2/3 phosphorylation.
  • Regulates AP-1 and ASK1-MAPK signaling pathways.
  • Inhibits ROS production.
  • Protects human keratinocytes against UVB-induced damage.
  • Applicable for research related to skin aging.