Learn More
Medchemexpress LLC (+)-Columbianetin acetate | 23180-65-6 | 288.30 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN0363A5MG
Encompass_Preferred
(S)-Columbianetin acetate is an isomer of Columbianetin, a phytoalexin associated with celery (Apium graveolens) resistance to pathogens. It exhibits notable anti-fungal and anti-inflammatory activity.
  • Isomer of columbianetin
  • Associated with celery's resistance to pathogens
  • Exhibits anti-fungal activity
  • Exhibits anti-inflammatory activity
  • For research use only
Catalog No. 50-003-88877