Learn More
TARGETMOL CHEMICALS INC COLUMBIN 50MG
SDP
Catalog No. 502576723
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. Columbin has anti-inflammatory activity. 2. Columbin has chemopreventive ability against human colon cancer. 3. Columbin inhibits PLA2 hydrolysis of ghost RBC in a dose-dependent fashion. purity: 99%
Catalog No. 50-257-6723 Supplier TARGETMOL CHEMICALS INC Supplier No. T4S059050MG
Also available in 5 mg 10 mg 25 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. Columbin has anti-inflammatory activity. 2. Columbin has chemopreventive ability against human colon cancer. 3. Columbin inhibits PLA2 hydrolysis of ghost RBC in a dose-dependent fashion. purity: 99%