Learn More
Cayman Chemical Columbianadin, Purity: ≥98%, CAS: 5058-13-9, 10 mg
SDP
Catalog No. 502939818
Encompass_Preferred
A coumarin with diverse biological activities; inhibits depolarization-induced calcium uptake in GH3 rat pituitary cells; inhibits HIV-1 replication in H9 cells (EC50 = 4.6 µM); inhibits IL-1β-induced production of IL-6 in A549 cells and reduces iNOS levels and LPS-induced NO production in MH-S cells; decreases the number of alveolar and interstitial macrophages and alveolar wall thickness in a mouse model of LPS-induced lung inflammation from 20-60 mg/kg; reduces formalin-induced paw licking in mice at 10 mg/kg
Catalog No. 50-293-9818 Supplier Cayman Chemical Supplier No. 2766110 MG
A coumarin with diverse biological activities; inhibits depolarization-induced calcium uptake in GH3 rat pituitary cells; inhibits HIV-1 replication in H9 cells (EC50 = 4.6 µM); inhibits IL-1β-induced production of IL-6 in A549 cells and reduces iNOS levels and LPS-induced NO production in MH-S cells; decreases the number of alveolar and interstitial macrophages and alveolar wall thickness in a mouse model of LPS-induced lung inflammation from 20-60 mg/kg; reduces formalin-induced paw licking in mice at 10 mg/kg