Learn More
MSC Columbus McKinnon Anchor Shackle, Round Pin, Carbon Steel, Galvanized, 1000 lb
Catalog No. 5000514141
Encompass
This anchor shackle features a round pin and is designed for various lifting and rigging applications. Constructed from carbon steel with a galvanized finish, it offers durability and corrosion resistance for demanding environments.
  • Features a round pin for secure connections
  • Made from durable carbon steel
  • Galvanized finish for corrosion resistance
  • 1000 lb working load limit for reliable performance
  • Meets ASTM A153 standards for quality assurance
  • Suitable for a wide range of industrial applications
Catalog No. 50-005-14141 Supplier MSC Supplier No. 64618374
This anchor shackle features a round pin and is designed for various lifting and rigging applications. Constructed from carbon steel with a galvanized finish, it offers durability and corrosion resistance for demanding environments.
  • Features a round pin for secure connections
  • Made from durable carbon steel
  • Galvanized finish for corrosion resistance
  • 1000 lb working load limit for reliable performance
  • Meets ASTM A153 standards for quality assurance
  • Suitable for a wide range of industrial applications