Learn More
MSC Columbus McKinnon Chain Shackle, Round Pin, 1/4 in, 1500 Lb Load Capacity, Galvanized
Catalog No. 5000530630
Encompass
The Columbus McKinnon M546G is a round pin chain shackle constructed from carbon steel with a galvanized finish. It is engineered for various lifting applications and has a working load limit of 1,500 Lb.
  • Made from technically advanced micro alloy steel with optimal hardness for strength and ductility.
  • Shackles show major deformation before failure, enhancing safety.
  • Working load limit and traceability codes are permanently marked on the body.
  • Features alloy quenched and tempered pins.
  • Round pin shackles have a 5:1 design factor.
  • Carbon-type shackles are 17% to 50% stronger than comparable-sized carbon shackles.
  • Chain shackles are best suited for straight-line pulls.
  • Meets dimensional requirements and exceeds performance requirements of RR-C-271.
  • Adheres to ASME B30.26 standards.
Catalog No. 50-005-30630 Supplier MSC Supplier No. 64618531
The Columbus McKinnon M546G is a round pin chain shackle constructed from carbon steel with a galvanized finish. It is engineered for various lifting applications and has a working load limit of 1,500 Lb.
  • Made from technically advanced micro alloy steel with optimal hardness for strength and ductility.
  • Shackles show major deformation before failure, enhancing safety.
  • Working load limit and traceability codes are permanently marked on the body.
  • Features alloy quenched and tempered pins.
  • Round pin shackles have a 5:1 design factor.
  • Carbon-type shackles are 17% to 50% stronger than comparable-sized carbon shackles.
  • Chain shackles are best suited for straight-line pulls.
  • Meets dimensional requirements and exceeds performance requirements of RR-C-271.
  • Adheres to ASME B30.26 standards.