Learn More
MSC Columbus McKinnon Hand Chain Hoist, 1 ton Capacity, 10 ft Lift, Hook Mount
Catalog No. 5000531380
Encompass
The CM Cyclone Hook Mount Hand Chain Hoist is renowned for its superior engineering, efficiency, and durability. This reliable hoist, manufactured in the USA with a lifetime warranty, features a 1 ton (2000 lb) capacity and a 10 ft lift, making it suitable for various lifting applications with its hook mount design.
  • Superior engineering, efficiency, and durability
  • Reliable design
  • Easy maintenance with interchangeable parts
  • Meets or exceeds HMI and ASME/ANSI performance and safety standards
  • Positive load control with enclosed automatic brake
  • High-strength aluminum alloy castings
  • Rugged alloy chain for enhanced strength
  • High-efficiency spur gearing for increased lift
  • Fully machined, forged lift wheel pockets
  • Made in the USA
  • Lifetime warranty
  • Metric rated
  • Integrated overload protection
  • Minimum headroom: 14.0000 in
Catalog No. 50-005-31380 Supplier MSC Supplier No. 89783914
The CM Cyclone Hook Mount Hand Chain Hoist is renowned for its superior engineering, efficiency, and durability. This reliable hoist, manufactured in the USA with a lifetime warranty, features a 1 ton (2000 lb) capacity and a 10 ft lift, making it suitable for various lifting applications with its hook mount design.
  • Superior engineering, efficiency, and durability
  • Reliable design
  • Easy maintenance with interchangeable parts
  • Meets or exceeds HMI and ASME/ANSI performance and safety standards
  • Positive load control with enclosed automatic brake
  • High-strength aluminum alloy castings
  • Rugged alloy chain for enhanced strength
  • High-efficiency spur gearing for increased lift
  • Fully machined, forged lift wheel pockets
  • Made in the USA
  • Lifetime warranty
  • Metric rated
  • Integrated overload protection
  • Minimum headroom: 14.0000 in