Learn More
MSC Columbus McKinnon Plain Trolley, Series 635, 2-Ton, Low Headroom, Ball Bearing
Catalog No. 5000515527
Encompass
The Columbus McKinnon Series 635 Plain Trolley is a low-headroom model designed for efficient and reliable material handling. Featuring a 2-ton capacity and lifetime lubricated double row ball bearings, it ensures high performance and minimal maintenance in various industrial applications.
  • Lifetime lubricated double row ball bearings for high efficiency, low maintenance.
  • Low headroom design.
  • Fits wide range of beam sizes for maximum versatility.
  • Steel plate side frames.
  • Steel spur gearing for optimum strength and smooth operation.
  • Universal tread hardened trackwheels that operate on standard S-beams or flat flanged beams.
  • Metric rated.
  • Rigged lug suspension (required for attaching to hoist, not included).
Catalog No. 50-005-15527 Supplier MSC Supplier No. 89817720
The Columbus McKinnon Series 635 Plain Trolley is a low-headroom model designed for efficient and reliable material handling. Featuring a 2-ton capacity and lifetime lubricated double row ball bearings, it ensures high performance and minimal maintenance in various industrial applications.
  • Lifetime lubricated double row ball bearings for high efficiency, low maintenance.
  • Low headroom design.
  • Fits wide range of beam sizes for maximum versatility.
  • Steel plate side frames.
  • Steel spur gearing for optimum strength and smooth operation.
  • Universal tread hardened trackwheels that operate on standard S-beams or flat flanged beams.
  • Metric rated.
  • Rigged lug suspension (required for attaching to hoist, not included).