Learn More
Waters Corp Column, 1.7 μm, 4.6 × 150 mm, Insulin
Catalog No. NC3936319 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The ACQUITY BEH125 SEC Column is designed for size-exclusion chromatography of insulin and related biomolecules. Built on BEH particle technology, this column delivers high efficiency and reproducible separation performance suitable for biomolecular characterization and quality assessment workflows involving insulin.
  • Designed for size-exclusion chromatography of insulin and related biomolecules.
  • Built on BEH particle technology.
  • Delivers high efficiency.
  • Offers reproducible separation performance.
  • Suitable for biomolecular characterization workflows.
  • Suitable for quality assessment workflows involving insulin.
Catalog No. NC3936319 Supplier Waters Corp Supplier No. 186008654
The ACQUITY BEH125 SEC Column is designed for size-exclusion chromatography of insulin and related biomolecules. Built on BEH particle technology, this column delivers high efficiency and reproducible separation performance suitable for biomolecular characterization and quality assessment workflows involving insulin.
  • Designed for size-exclusion chromatography of insulin and related biomolecules.
  • Built on BEH particle technology.
  • Delivers high efficiency.
  • Offers reproducible separation performance.
  • Suitable for biomolecular characterization workflows.
  • Suitable for quality assessment workflows involving insulin.