Learn More
Ace Glass, Inc. 10mm ID 18" 35/20 epoxy coated flash chromatography column with porosity B filter disc.
SDP
Catalog No. 50972495
Encompass_Preferred
10mm I.D., 8 simple absorption flash chromatography column for rapid preparative separations with moderate resolution. Allows separations of samples from 0.01 to 10.0g in 10-15 minutes. Column is epoxy coated and has 35/20 joints. Includes porosity B filter disc.
Catalog No. 50-972-495 Supplier Ace Glass, Inc. Supplier No. 587250
10mm I.D., 8 simple absorption flash chromatography column for rapid preparative separations with moderate resolution. Allows separations of samples from 0.01 to 10.0g in 10-15 minutes. Column is epoxy coated and has 35/20 joints. Includes porosity B filter disc.