Learn More
Ace Glass, Inc. 10mm ID 18" 35/20 epoxy coated flash chromatography column.
SDP
Catalog No. 50972466
Encompass_Preferred
10mm I.D., 18 simple absorption flash chromatography column for rapid preparative separations with moderate resolution. Allows separations of samples from 0.01 to 10.0g in 10-15 minutes. Column is epoxy coated and has 35/20 joints.
Catalog No. 50-972-466 Supplier Ace Glass, Inc. Supplier No. 587206
10mm I.D., 18 simple absorption flash chromatography column for rapid preparative separations with moderate resolution. Allows separations of samples from 0.01 to 10.0g in 10-15 minutes. Column is epoxy coated and has 35/20 joints.