Learn More
Ace Glass, Inc. 13mm ID 10" 35/20 epoxy coated flash chromatography column with porosity B filter disc.
SDP
Catalog No. 50978573
Encompass_Preferred
13mm I.D., 10 simple absorption flash chromatography column for rapid preparative separations with moderate resolution. Allows separations of samples from 0.01 to 10.0g in 10-15 minutes. Column is epoxy coated and has 35/20 joints. Includes porosity B filter disc.
Catalog No. 50-978-573 Supplier Ace Glass, Inc. Supplier No. 587257
13mm I.D., 10 simple absorption flash chromatography column for rapid preparative separations with moderate resolution. Allows separations of samples from 0.01 to 10.0g in 10-15 minutes. Column is epoxy coated and has 35/20 joints. Includes porosity B filter disc.