Learn More
Ace Glass, Inc. 13mm ID 10" 35/20 epoxy coated flash chromatography column.
SDP
Catalog No. 50972464
Encompass_Preferred
13mm I.D., 10 simple absorption flash chromatography column for rapid preparative separations with moderate resolution. Allows separations of samples from 0.01 to 10.0g in 10-15 minutes. Column is epoxy coated and has 35/20 joints.
Catalog No. 50-972-464 Supplier Ace Glass, Inc. Supplier No. 587203
13mm I.D., 10 simple absorption flash chromatography column for rapid preparative separations with moderate resolution. Allows separations of samples from 0.01 to 10.0g in 10-15 minutes. Column is epoxy coated and has 35/20 joints.