Learn More
Ace Glass, Inc. 13mm ID 12" 24/40 epoxy coated flash chromatography column with porosity B filter disc.
SDP
Catalog No. NC0790200
Encompass_Preferred
13mm I.D. x 305mm long, simple absorption flash chromatography column for rapid preparative separations. Column is epoxy coated for safety, and has 70-100 micron porosity sealed-in fritted disc above the 2mm bore PTFE stopcock. 40mL capacity.
Catalog No. NC0790200 Supplier Ace Glass, Inc. Supplier No. 587155
13mm I.D. x 305mm long, simple absorption flash chromatography column for rapid preparative separations. Column is epoxy coated for safety, and has 70-100 micron porosity sealed-in fritted disc above the 2mm bore PTFE stopcock. 40mL capacity.