Learn More
Ace Glass, Inc. 13mm ID 12" 35/20 epoxy coated flash chromatography column.
SDP
Catalog No. 50978561
Encompass_Preferred
13mm I.D., 12 simple absorption flash chromatography column for rapid preparative separations with moderate resolution. Allows separations of samples from 0.01 to 10.0g in 10-15 minutes. Column is epoxy coated and has 35/20 joints.
Catalog No. 50-978-561 Supplier Ace Glass, Inc. Supplier No. 587204
13mm I.D., 12 simple absorption flash chromatography column for rapid preparative separations with moderate resolution. Allows separations of samples from 0.01 to 10.0g in 10-15 minutes. Column is epoxy coated and has 35/20 joints.