Learn More
Ace Glass, Inc. 13mm ID 18" 35/20 epoxy coated flash chromatography column.
SDP
Catalog No. 50972465
Encompass_Preferred
13mm I.D., 18 simple absorption flash chromatography column for rapid preparative separations with moderate resolution. Allows separations of samples from 0.01 to 10.0g in 10-15 minutes. Column is epoxy coated and has 35/20 joints.
Catalog No. 50-972-465 Supplier Ace Glass, Inc. Supplier No. 587205
13mm I.D., 18 simple absorption flash chromatography column for rapid preparative separations with moderate resolution. Allows separations of samples from 0.01 to 10.0g in 10-15 minutes. Column is epoxy coated and has 35/20 joints.