Learn More
Ace Glass, Inc. 13mm ID 8" 35/20 epoxy coated flash chromatography column with porosity B filter disc.
SDP
Catalog No. 50972499
Encompass_Preferred
13mm I.D., 8 simple absorption flash chromatography column for rapid preparative separations with moderate resolution. Allows separations of samples from 0.01 to 10.0g in 10-15 minutes. Column is epoxy coated and has 35/20 joints. Includes porosity B filter disc.
Catalog No. 50-972-499 Supplier Ace Glass, Inc. Supplier No. 587255
13mm I.D., 8 simple absorption flash chromatography column for rapid preparative separations with moderate resolution. Allows separations of samples from 0.01 to 10.0g in 10-15 minutes. Column is epoxy coated and has 35/20 joints. Includes porosity B filter disc.